Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710239.3 | XP_002710285.3 | 2 | 1211 | 95 | |
GenBank mRNA | AJ291320.1 | - | 1 | 252 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |