Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163067.1 | NP_001156539.1 | 4 - 5 | 649 | 86 | |
XM_008274050.2 | XP_008272272.1 | 4 - 5 | 638 | 86 | ||
XM_008274053.2 | XP_008272275.1 | 4 - 5 | 427 | 86 | ||
XM_008274060.2 | XP_008272282.1 | 4 - 5 | 540 | 86 | ||
XM_017350167.1 | XP_017205656.1 | 5 - 6 | 639 | 86 | ||
GenBank mRNA | AY165034.1 | - | 4 - 5 | 649 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |