Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082165.1 | NP_001075634.1 | 2 - 3 | 214 | 66 | |
GenBank mRNA | AY471576.1 | - | 2 - 3 | 214 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |