Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082007.1 | NP_001075476.1 | 1 - 2 | 61 | 70 | |
XM_008263944.2 | XP_008262166.1 | 6 - 7 | 677 | 70 | ||
GenBank mRNA | AB042999.1 | - | 1 - 2 | 61 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |