Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPY4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160280.1 | NP_001153752.1 | 1 | 750 | 93 | |
XM_008269879.2 | XP_008268101.1 | 2 | 906 | 93 | ||
GenBank mRNA | AB469825.1 | - | 1 | 750 | 93 | |
AY587060.1 | - | 1 | 393 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Oryctolagus cuniculus Both primers and probe map within a single exon |