Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259599.2 | XP_008257821.1 | 14 - 15 | 2731 | 94 | |
XM_017343367.1 | XP_017198856.1 | 14 - 15 | 2095 | 94 | ||
GenBank mRNA | AF184165.1 | - | 3 - 4 | 197 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |