Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL10A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714724.3 | XP_002714770.1 | 3 | 386 | 98 | |
GenBank mRNA | AF247705.1 | - | 2 | 154 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |