Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168395.1 | NP_001161867.1 | 515 | 78 | ||
GenBank mRNA | AB008784.1 | - | 515 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus |