Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168397.1 | NP_001161869.1 | 357 | 55 | ||
GenBank mRNA | AB008785.1 | - | 357 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus |