Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082015.1 | NP_001075484.1 | 927 | 69 | ||
GenBank mRNA | AB231473.1 | - | 927 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus |