Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170871.1 | NP_001164342.1 | 8 - 9 | 1262 | 77 | |
GenBank mRNA | AJ002388.1 | - | 8 - 9 | 1262 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Probe spans exons |