Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171017.1 | NP_001164488.1 | 5 - 6 | 1071 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |