Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171082.1 | NP_001164553.1 | 1 | 552 | 63 | |
XM_017342635.1 | XP_017198124.1 | 2 | 785 | 63 | ||
GenBank mRNA | M58021.1 | - | 1 | 563 | 63 | |
M82873.1 | - | 1 | 552 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |