Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171113.1 | NP_001164584.1 | 2 - 3 | 390 | 75 | |
GenBank mRNA | K01521.1 | - | 2 - 3 | 301 | 75 | |
M19137.1 | - | 2 - 3 | 390 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |