Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171274.1 | NP_001164745.1 | 3 - 4 | 763 | 83 | |
XM_017344299.1 | XP_017199788.1 | 4 - 5 | 893 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |