Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171276.1 | NP_001164747.1 | 1 | 642 | 67 | |
XM_008261492.2 | XP_008259714.1 | 2 | 765 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon |