Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171283.1 | NP_001164754.1 | 1 - 2 | 165 | 103 | |
XM_008261382.2 | XP_008259604.1 | 2 - 3 | 249 | 103 | ||
XM_008261383.2 | XP_008259605.1 | 2 - 3 | 312 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |