Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171298.1 | NP_001164769.1 | 2 - 3 | 120 | 57 | |
XM_008264020.2 | XP_008262242.1 | 2 - 3 | 88 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |