Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171298.1 | NP_001164769.1 | 3 - 4 | 209 | 87 | |
XM_008264020.2 | XP_008262242.1 | 3 - 4 | 177 | 87 | ||
XM_008264021.2 | XP_008262243.1 | 2 - 3 | 337 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |