Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171305.1 | NP_001164776.1 | 1 - 2 | 141 | 78 | |
XM_008264042.2 | XP_008262264.1 | 1 - 2 | 145 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |