Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPIFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171359.1 | NP_001164830.1 | 5 - 6 | 585 | 75 | |
XM_008257060.2 | XP_008255282.1 | 6 - 7 | 693 | 75 | ||
XM_008257061.2 | XP_008255283.1 | 7 - 8 | 2039 | 75 | ||
XM_008257062.2 | XP_008255284.1 | 5 - 6 | 565 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |