Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171362.1 | NP_001164833.1 | 1490 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |