Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171382.1 | NP_001164853.1 | 1294 | 73 | ||
XM_008250285.2 | XP_008248507.1 | 1567 | 73 | |||
XM_008250286.2 | XP_008248508.1 | 1450 | 73 | |||
XM_008250287.2 | XP_008248509.1 | 1352 | 73 | |||
XM_008250288.2 | XP_008248510.1 | 1451 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus |