Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171405.1 | NP_001164876.1 | 2 - 3 | 684 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |