Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171497.1 | NP_001164968.1 | 1 - 2 | 478 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons |