Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195764.1 | NP_001182693.1 | 342 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus |