Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205004.1 | NP_001191933.1 | 1 - 2 | 345 | 70 | |
XM_017344516.1 | XP_017200005.1 | 1 - 2 | 439 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |