Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032840.1 | NP_001028012.1 | 122 | 56 | ||
GenBank mRNA | AF345333.1 | - | 122 | 56 | ||
JU332423.1 | - | 73 | 56 | |||
JU332424.1 | - | 73 | 56 | |||
JV045848.1 | - | 169 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |