Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193903.2 | NP_001180832.1 | 869 | 93 | ||
XM_028840457.1 | XP_028696290.1 | 868 | 93 | |||
XM_028840458.1 | XP_028696291.1 | 790 | 93 | |||
XM_028840459.1 | XP_028696292.1 | 868 | 93 | |||
XM_028840461.1 | XP_028696294.1 | 868 | 93 | |||
XM_028840462.1 | XP_028696295.1 | 868 | 93 | |||
XM_028840463.1 | XP_028696296.1 | 868 | 93 | |||
XM_028840464.1 | XP_028696297.1 | 868 | 93 | |||
XM_028840465.1 | XP_028696298.1 | 868 | 93 | |||
GenBank mRNA | JV647373.1 | - | 756 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |