Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194354.2 | NP_001181283.1 | 534 | 76 | ||
XM_015134931.2 | XP_014990417.1 | 1000 | 76 | |||
GenBank mRNA | JU321403.1 | - | 818 | 76 | ||
JV045303.1 | - | 664 | 76 | |||
JV588958.1 | - | 801 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |