Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082002.4 | XP_001082002.3 | 1026 | 88 | ||
GenBank mRNA | JU321906.1 | - | 138 | 88 | ||
JU477206.1 | - | 151 | 88 | |||
JV048254.1 | - | 161 | 88 | |||
JV661400.1 | - | 151 | 88 | |||
JV668935.1 | - | 211 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |