Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098374.4 | XP_001098374.1 | 350 | 63 | ||
GenBank mRNA | JV463266.1 | - | 68 | 63 | ||
JV611756.1 | - | 335 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |