Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115825.2 | XP_014971311.2 | 3169 | 133 | ||
XM_015115826.2 | XP_014971312.2 | 3169 | 133 | |||
XM_028834035.1 | XP_028689868.1 | 3266 | 133 | |||
XM_028834036.1 | XP_028689869.1 | 653 | 133 | |||
GenBank mRNA | JU343756.1 | - | 2733 | 133 | ||
JU486874.1 | - | 2745 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |