Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092459.4 | XP_001092459.1 | 462 | 84 | ||
XM_028845935.1 | XP_028701768.1 | 1129 | 84 | |||
XM_028845936.1 | XP_028701769.1 | 982 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |