Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097187.4 | XP_001097187.1 | 313 | 71 | ||
XM_015115483.2 | XP_014970969.1 | 307 | 71 | |||
GenBank mRNA | JU326704.1 | - | 256 | 71 | ||
JV710422.1 | - | 259 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |