Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257637.1 | NP_001244566.1 | 339 | 124 | ||
XM_015133044.2 | XP_014988530.2 | 531 | 124 | |||
XM_015133045.2 | XP_014988531.2 | 431 | 124 | |||
XM_015133046.2 | XP_014988532.2 | 1611 | 124 | |||
XM_015133047.2 | XP_014988533.2 | 430 | 124 | |||
XM_028844937.1 | XP_028700770.1 | 535 | 124 | |||
GenBank mRNA | JU335567.1 | - | 339 | 124 | ||
JU335568.1 | - | 339 | 124 | |||
JU476486.1 | - | 442 | 124 | |||
JU476487.1 | - | 442 | 124 | |||
JV044242.1 | - | 469 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |