Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266369.1 | NP_001253298.1 | 1075 | 144 | ||
XM_015145659.2 | XP_015001145.1 | 1105 | 144 | |||
XM_015145660.2 | XP_015001146.1 | 1105 | 144 | |||
XM_028852185.1 | XP_028708018.1 | 1105 | 144 | |||
XM_028852187.1 | XP_028708020.1 | 1131 | 144 | |||
GenBank mRNA | JU321225.1 | - | 1006 | 144 | ||
JU321226.1 | - | 1006 | 144 | |||
JU321229.1 | - | 1006 | 144 | |||
JU470512.1 | - | 1076 | 144 | |||
JV044132.1 | - | 1075 | 144 | |||
JV633395.1 | - | 1078 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |