Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266881.1 | NP_001253810.1 | 146 | 113 | ||
GenBank mRNA | JU335507.1 | - | 146 | 113 | ||
JV528251.1 | - | 148 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |