Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194551.2 | NP_001181480.2 | 916 | 63 | ||
XM_015126676.2 | XP_014982162.2 | 848 | 63 | |||
XM_015126677.2 | XP_014982163.2 | 891 | 63 | |||
GenBank mRNA | JU336620.1 | - | 822 | 63 | ||
JU336621.1 | - | 822 | 63 | |||
JU336622.1 | - | 822 | 63 | |||
JU336623.1 | - | 822 | 63 | |||
JU336624.1 | - | 822 | 63 | |||
JU336625.1 | - | 822 | 63 | |||
JU336626.1 | - | 822 | 63 | |||
JU471395.1 | - | 796 | 63 | |||
JU471396.1 | - | 796 | 63 | |||
JU471397.1 | - | 796 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |