Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126323.2 | XP_014981809.1 | 782 | 79 | ||
XM_028840380.1 | XP_028696213.1 | 665 | 79 | |||
GenBank mRNA | JU336882.1 | - | 618 | 79 | ||
JU472750.1 | - | 699 | 79 | |||
JV043620.1 | - | 741 | 79 | |||
JV664317.1 | - | 744 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |