Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808387.3 | XP_002808433.1 | 901 | 96 | ||
GenBank mRNA | JU487310.1 | - | 576 | 96 | ||
JV617733.1 | - | 685 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |