Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1I1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261582.1 | NP_001248511.1 | 2014 | 69 | ||
XM_015134250.2 | XP_014989736.1 | 2159 | 69 | |||
XM_015134251.2 | XP_014989737.1 | 1957 | 69 | |||
XM_015134253.2 | XP_014989739.1 | 1906 | 69 | |||
XM_015134255.2 | XP_014989741.1 | 2099 | 69 | |||
XM_015134256.2 | XP_014989742.1 | 2048 | 69 | |||
XM_028844745.1 | XP_028700578.1 | 2012 | 69 | |||
XM_028844746.1 | XP_028700579.1 | 1961 | 69 | |||
GenBank mRNA | AY369807.1 | - | 274 | 69 | ||
JU319819.1 | - | 2014 | 69 | |||
JV566152.1 | - | 2039 | 69 | |||
JV703812.1 | - | 1996 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |