Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257767.1 | NP_001244696.1 | 626 | 95 | ||
GenBank mRNA | JU319824.1 | - | 626 | 95 | ||
JU473388.1 | - | 518 | 95 | |||
JV045780.1 | - | 599 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |