Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194661.1 | NP_001181590.1 | 362 | 79 | ||
GenBank mRNA | JU326882.1 | - | 305 | 79 | ||
JV046211.1 | - | 351 | 79 | |||
JV642420.1 | - | 357 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |