Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266428.1 | NP_001253357.1 | 235 | 93 | ||
GenBank mRNA | JU321845.1 | - | 300 | 93 | ||
JU477540.1 | - | 235 | 93 | |||
JV048961.1 | - | 307 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |