Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266917.1 | NP_001253846.1 | 246 | 65 | ||
XM_028850456.1 | XP_028706289.1 | 1489 | 65 | |||
XM_028850457.1 | XP_028706290.1 | 485 | 65 | |||
XM_028850458.1 | XP_028706291.1 | 248 | 65 | |||
XM_028850459.1 | XP_028706292.1 | 138 | 65 | |||
GenBank mRNA | JU321454.1 | - | 246 | 65 | ||
JU335545.1 | - | 333 | 65 | |||
JV047710.1 | - | 424 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |