Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261065.1 | NP_001247994.1 | 1395 | 106 | ||
XM_015151639.2 | XP_015007125.1 | 3208 | 106 | |||
GenBank mRNA | JU334515.1 | - | 1395 | 106 | ||
JV684155.1 | - | 1422 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |