Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148701.2 | XP_015004187.1 | 274 | 128 | ||
XM_028828541.1 | XP_028684374.1 | 274 | 128 | |||
GenBank mRNA | JU487950.1 | - | 204 | 128 | ||
JV666253.1 | - | 227 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |