Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194429.3 | NP_001181358.1 | 156 | 99 | ||
GenBank mRNA | JU325979.1 | - | 121 | 99 | ||
JU325980.1 | - | 121 | 99 | |||
JV704485.1 | - | 134 | 99 | |||
JV704486.1 | - | 134 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |