Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148649.2 | XP_015004135.1 | 198 | 62 | ||
XM_015148650.2 | XP_015004136.2 | 198 | 62 | |||
XM_028828701.1 | XP_028684534.1 | 212 | 62 | |||
GenBank mRNA | JV527813.1 | - | 143 | 62 | ||
JV625116.1 | - | 110 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |